ENA429360529 Enamine

2-(4-iodophenoxy)acetonitrile

  • MDL number MFCD11176886
  • Cas Number : 243657-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-57
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190074
Availability details for
Quantity
  • Availability
  • Add to Cart