ENA429360532 Enamine

{3-[(2,3-dihydro-1H-inden-2-yl)amino]phenyl}methanol

  • MDL number MFCD12037074
  • Cas Number : 1157346-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190077
Availability details for
Quantity
  • Availability
  • Add to Cart