ENA429360534 Enamine

1-(1,3-dimethyl-1H-indol-2-yl)methanamine

  • MDL number MFCD20717950
  • Cas Number : 169615-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190079
Availability details for
Quantity
  • Availability
  • Add to Cart