ENA429360536 Enamine

5-methoxy-2-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD09880307
  • Cas Number : 861374-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189490

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189490
Availability details for
Quantity
  • Availability
  • Add to Cart