ENA429360537 Enamine

1-methanesulfonyl-3,3-dimethylpiperazine

  • MDL number MFCD19285581
  • Cas Number : 141923-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190083

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190083
Availability details for
Quantity
  • Availability
  • Add to Cart