ENA429360540 Enamine

3-hydroxypentanedioic acid

  • MDL number MFCD06208235
  • Cas Number : 638-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190087

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190087
Availability details for
Quantity
  • Availability
  • Add to Cart