ENA429360543 Enamine

methyl 2-(1-aminocyclopentyl)acetate hydrochloride

  • MDL number MFCD28125059
  • Cas Number : 1224474-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190092
Availability details for
Quantity
  • Availability
  • Add to Cart