ENA429360550 Enamine

N,3-dimethylpyridin-2-amine

  • MDL number MFCD16697578
  • Cas Number : 156267-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190110
Availability details for
Quantity
  • Availability
  • Add to Cart