ENA429360553 Enamine

4-(2-bromoethyl)pyridine hydrobromide

  • MDL number MFCD12031796
  • Cas Number : 120277-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190131

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-190131
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart