ENA429360557 Enamine

3-acetamido-2,2-dimethylpropanoic acid

  • MDL number MFCD20644782
  • Cas Number : 80253-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189495
Availability details for
Quantity
  • Availability
  • Add to Cart