ENA429360562 Enamine

5-(3-fluoropyridin-4-yl)-1,2,4-oxadiazol-3-amine

  • MDL number MFCD21115083
  • Cas Number : 1490982-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189500

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189500
Availability details for
Quantity
  • Availability
  • Add to Cart