ENA429360566 Enamine

1-(3-amino-4-chlorophenyl)propan-1-one

  • MDL number MFCD11935014
  • Cas Number : 56721-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189504
Availability details for
Quantity
  • Availability
  • Add to Cart