ENA429360572 Enamine

4-[(2R,6S)-2,6-dimethylmorpholin-4-yl]aniline

  • MDL number MFCD24646311
  • Cas Number : 342578-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190182
Availability details for
Quantity
  • Availability
  • Add to Cart