ENA429360574 Enamine

3-(5-amino-2-fluorophenyl)-1,3-oxazolidin-2-one

  • MDL number MFCD11167816
  • Cas Number : 1038282-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190202

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190202
Availability details for
Quantity
  • Availability
  • Add to Cart