ENA429360575 Enamine

2-[(5-bromo-2-nitrophenyl)amino]-2-methylpropanoic acid

  • MDL number MFCD21513350
  • Cas Number : 1491844-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189505
Availability details for
Quantity
  • Availability
  • Add to Cart