ENA429360577 Enamine

methyl 4-(chloromethyl)-3-fluorobenzoate

  • MDL number MFCD26863449
  • Cas Number : 1567097-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190209
Availability details for
Quantity
  • Availability
  • Add to Cart