ENA429360579 Enamine

1-(3-aminophenyl)-1,3,3-trimethylurea

  • MDL number MFCD21809520
  • Cas Number : 1505050-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190216
Availability details for
Quantity
  • Availability
  • Add to Cart