ENA429360580 Enamine

2-(aminomethyl)-4-methylpentan-1-ol

  • MDL number MFCD16706733
  • Cas Number : 1250484-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190220

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190220
Availability details for
Quantity
  • Availability
  • Add to Cart