ENA429360583 Enamine

2-{1,4-dioxaspiro[4.5]decan-8-ylidene}acetonitrile

  • MDL number MFCD11847777
  • Cas Number : 124499-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190223
Availability details for
Quantity
  • Availability
  • Add to Cart