ENA429360584 Enamine

N-{1-azabicyclo[3.2.1]octan-4-ylidene}hydroxylamine

  • Cas Number : 1026035-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190225
Availability details for
Quantity
  • Availability
  • Add to Cart