ENA429360587 Enamine

5-(5-fluoro-2-methylphenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD13873747
  • Cas Number : 299441-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190237
Availability details for
Quantity
  • Availability
  • Add to Cart