ENA429360588 Enamine

5-(4-chloro-2-fluorophenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD11137530
  • Cas Number : 635301-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190238
Availability details for
Quantity
  • Availability
  • Add to Cart