ENA429360589 Enamine

N-[3-(hydroxymethyl)phenyl]propanamide

  • MDL number MFCD12818537
  • Cas Number : 81863-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190240

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190240
Availability details for
Quantity
  • Availability
  • Add to Cart