ENA429360591 Enamine

1,2,3,5-tetrahydro-4,1-benzoxazepine

  • MDL number MFCD11036257

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190243
Availability details for
Quantity
  • Availability
  • Add to Cart