ENA429360592 Enamine

2-cyano-4-(trifluoromethyl)benzene-1-sulfonamide

  • MDL number MFCD16093727
  • Cas Number : 1248024-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189457

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189457
Availability details for
Quantity
  • Availability
  • Add to Cart