ENA429360596 Enamine

4-(3,4-dichlorophenyl)but-3-en-2-one

  • MDL number MFCD00052849
  • Cas Number : 74546-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190248

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190248
Availability details for
Quantity
  • Availability
  • Add to Cart