ENA429360597 Enamine

3-{[(tert-butoxy)carbonyl]amino}-2-(4-fluorophenyl)propanoic acid

  • MDL number MFCD24531585
  • Cas Number : 1690999-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190249
Availability details for
Quantity
  • Availability
  • Add to Cart