ENA429360598 Enamine

1-[5-(3-fluoropyridin-4-yl)-1,3,4-thiadiazol-2-yl]methanamine

  • MDL number MFCD21289007
  • Cas Number : 1466227-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190252

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190252
Availability details for
Quantity
  • Availability
  • Add to Cart