ENA429360601 Enamine

ethyl 2-cyano-5-fluorobenzoate

  • MDL number MFCD11041402
  • Cas Number : 1260751-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190256
Availability details for
Quantity
  • Availability
  • Add to Cart