ENA429360602 Enamine

1-methyl-3-(oxan-3-yl)-1H-pyrazol-5-amine

  • MDL number MFCD17275110
  • Cas Number : 1339351-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189515
Availability details for
Quantity
  • Availability
  • Add to Cart