ENA429360605 Enamine

heptane-4-sulfonyl chloride

  • MDL number MFCD16671785
  • Cas Number : 920-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190261
Availability details for
Quantity
  • Availability
  • Add to Cart