ENA429360606 Enamine

(2-amino-3,6-dichlorophenyl)methanol

  • MDL number MFCD20695928
  • Cas Number : 58579-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190262
Availability details for
Quantity
  • Availability
  • Add to Cart