ENA429360607 Enamine

4-(1-methyl-1H-pyrazol-4-yl)benzaldehyde

  • MDL number MFCD08669908
  • Cas Number : 179055-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190263
Availability details for
Quantity
  • Availability
  • Add to Cart