ENA429360609 Enamine

5-(1H-imidazol-2-yl)-2-methylaniline

  • MDL number MFCD08668915
  • Cas Number : 1126367-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190265
Availability details for
Quantity
  • Availability
  • Add to Cart