ENA429360612 Enamine

1,3-oxathiolan-2-one

  • MDL number MFCD06797711
  • Cas Number : 3326-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190268
Availability details for
Quantity
  • Availability
  • Add to Cart