ENA429360617 Enamine

3-bromobenzene-1,2-dicarbonitrile

  • MDL number MFCD19689407
  • Cas Number : 76241-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189516
Availability details for
Quantity
  • Availability
  • Add to Cart