ENA429360619 Enamine

5-bromo-1-methyl-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one

  • MDL number MFCD22394552
  • Cas Number : 175424-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189517

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189517
Availability details for
Quantity
  • Availability
  • Add to Cart