ENA429360622 Enamine

[1-(propan-2-yl)-1H-1,2,4-triazol-5-yl]methanamine

  • MDL number MFCD21269625
  • Cas Number : 1469241-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190285
Availability details for
Quantity
  • Availability
  • Add to Cart