ENA429360625 Enamine

[(2-chloroethanesulfonyl)methyl]benzene

  • MDL number MFCD00018967
  • Cas Number : 66998-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190291

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190291
Availability details for
Quantity
  • Availability
  • Add to Cart