ENA429360627 Enamine

[2-({8-oxa-3,5-diazatricyclo[7.4.0.0,2,7]trideca-1(9),2,4,6,10,12-hexaen-6-yl}amino)cyclohexyl]methanol

  • MDL number MFCD28125081
  • Cas Number : 1803591-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190294
Availability details for
Quantity
  • Availability
  • Add to Cart