ENA429360632 Enamine

4-chloro-6-fluoro-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD20485537
  • Cas Number : 940054-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190304
Availability details for
Quantity
  • Availability
  • Add to Cart