ENA429360633 Enamine

[1-(2-amino-6-chloropyrimidin-4-yl)pyrrolidin-2-yl]methanol

  • MDL number MFCD23722397
  • Cas Number : 1494756-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190322
Availability details for
Quantity
  • Availability
  • Add to Cart