ENA429360640 Enamine

4-(4-chlorophenyl)-4-methylpiperidine hydrochloride

  • MDL number MFCD14582953
  • Cas Number : 324002-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190340
Availability details for
Quantity
  • Availability
  • Add to Cart