ENA429360643 Enamine

3-[(1H-imidazol-1-yl)methyl]benzoic acid hydrochloride

  • MDL number MFCD08690248
  • Cas Number : 218131-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190349
Availability details for
Quantity
  • Availability
  • Add to Cart