ENA429360666 Enamine

2-[(4-fluorophenyl)amino]ethane-1-thiol

  • MDL number MFCD00022100
  • Cas Number : 20295-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190385
Availability details for
Quantity
  • Availability
  • Add to Cart