ENA429360667 Enamine

8-bromo-2-methyl-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD19377261
  • Cas Number : 868371-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190386
Availability details for
Quantity
  • Availability
  • Add to Cart