ENA429360668 Enamine

[1-(4-chlorophenyl)-2-ethylbutyl](methyl)amine

  • MDL number MFCD12182017
  • Cas Number : 1156263-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190387

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190387
Availability details for
Quantity
  • Availability
  • Add to Cart