ENA429360669 Enamine

1-(2-chloropyridin-4-yl)ethan-1-ol

  • MDL number MFCD12923076
  • Cas Number : 1245644-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190388
Availability details for
Quantity
  • Availability
  • Add to Cart