ENA429360675 Enamine

N-(4,6-dimethylpyrimidin-2-yl)prop-2-enamide

  • MDL number MFCD12091213
  • Cas Number : 1156159-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190400

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190400
Availability details for
Quantity
  • Availability
  • Add to Cart