ENA429360680 Enamine

1-methyl-2,3-dihydro-1H-indol-5-ol hydrobromide

  • MDL number MFCD28118842
  • Cas Number : 134328-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189528
Availability details for
Quantity
  • Availability
  • Add to Cart